Order Entry
Switzerland
ContactUsLinkComponent
Anti-ACTR1A Rabbit Polyclonal Antibody
Anti-ACTR1A Rabbit Polyclonal Antibody
Catalog # ABGEAP16117C
Supplier:  Abgent
Anti-ACTR1A Rabbit Polyclonal Antibody
Catalog # ABGEAP16117C
Supplier:  Abgent

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    ARP1 Actin-related Protein 1 Homolog A, Centractin alpha
  • Simbolo dell'antigene:
    ACTR1A
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Reattività:
    Mouse
  • Western blot:
    Yes
  • Epitopo:
    Center
  • ID gene:
    10121
  • Sinomini antigene:
    ARP1, ARP1 actin-related protein 1 homolog A, centractin alpha (yeast), actin-RPV, centractin alpha, ARP1, yeast homolog A, centrosome-associated actin homolog, ARP1 (actin-related protein 1, yeast) homolog A (centractin alpha), CTRN1, FLJ52695, FLJ52800, FLJ55002, ARP1 actin-related protein 1 homolog A, centractin alpha
  • Tampone di conservazione:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Peso molecolare:
    42.614 kDa
  • Sequenza:
    APLNPRKNRERAAEC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0,4 mg/ml
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This ACTR1A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 98-126 amino acids from the Central region of human ACTR1A.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ACTR1A (ARP1 actin-related protein 1 homolog A, centractin alpha (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse