Anti-MAP2K6 Rabbit Polyclonal Antibody
Catalog # ABGEAP16108A
Supplier: Abgent
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Mitogen-activated protein kinase Kinase 6
- Simbolo dell'antigene:MAP2K6
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- ImmunoChimica:Yes
- Reattività:Mouse
- Western blot:Yes
- Epitopo:N-Terminal
- ID gene:26399
- Sinomini antigene:PRKMK6|MKK6|MAPKK6|SAPKK-3|MEK6|SAPKK3
- Tampone di conservazione:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Peso molecolare:37.432 kDa
- Sequenza:NFEVKADDLEPIVC
- Temperatura di conservazione:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentrazione:0,5 mg/ml
- Temperatura di spedizione:4 °C
- Immunogeno:This Mouse Map2k6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 30-57 amino acids from the N-terminal region of mouse Map2k6.
- Purificazione:Protein A & Peptide affinity purification
- Conf:400 µl
Specifications
About this item
Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse