Order Entry
Switzerland
ContactUsLinkComponent
Anti-CASP6 Rabbit Polyclonal Antibody
Anti-CASP6 Rabbit Polyclonal Antibody
Catalog # ABGEAP1313B
Supplier:  Abgent
undefined
Anti-CASP6 Rabbit Polyclonal Antibody
Catalog # ABGEAP1313B
Supplier:  Abgent

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Caspase 6
  • Simbolo dell'antigene:
    CASP6
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Reattività:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitopo:
    C-terminal
  • Sinomini antigene:
    caspase 6|apoptotic protease MCH-2|caspase-6|CASP-6|apoptosis-related cysteine protease
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Conf:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CASP6 (caspase 6, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse