Anti-CSPS Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0219
Supplier: AbFrontier
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:6818
- Sinomini antigene:STM|TL-PST|HAST3|ST1A3|M-PST|HAST|ST1A3/ST1A4|ST1A5
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli
- Purificazione:purified from E.coli
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
The human Catecholamine-Sulfating Phenol Sulfotransferase (CSPS) is the only sulfotransferase that catalyses the sulfation of catecholamins, in particular the neurotransmitter dopamine, with high activity. CSPS is required for stimulation by Mn2+ of the sulfating activity and expressed in the human intestine, brain, platelet and other tissues. In the brain it may play a role in regulating the levels of dopamine. It also serves as a detoxifying function in the intestine, where it may detoxify potentially lethal dietary monoamines.
Western Blot: 1:500~2,000;
Type: Primary
Antigen: CSPS (catecholamine-sulfating phenol sulfotransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human