Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Tumor protein p53
- Simbolo dell'antigene:P53
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:7157
- Sinomini antigene:P53|BCC7|TRP53|LFS1
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Synthetic phospho-peptide
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
p53 is a transcription factor that regulates the cell cycle and hence functions as a tumor suppressor. p53 has been described as "the guardian of the genome", referring to its role in conserving stability by preventing genome mutation. p53 has many anti-cancer mechanisms: activating DNA repair proteins when DNA has sustained damage, holding the cell cycle at the G1/S regulation point on DNA damage recognition, initiating apoptosis if the DNA damage proves to be irrepairable. Human p53 is 393 amino acids long and has three domains: N-terminal transcription-activation domain (TAD), which activates transcription factors. 2) central DNA-binding core domain (DBD) 3) C-terminal homo-oligomerisation domain (OD); tetramerization greatly increases the activity of p53 in vivo. Mutations that deactivate p53 in cancer usually occur in the DBD and most of these mutations destroy the ability of the protein to bind to its target DNA sequences.
The activity of p53 is regulated at different levels and includes control by multi-site phosphorylation. p53 is phosphorylated (Ser6 and Ser9) by casein kinase 1δ and casein kinase 1ε both in vitro and in vivo.
Western Blot: 1:1,000;
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human