- Tipo anticorpo:Primary
- Tipo di anticorpi:microtubule-associated protein tau
- Simbolo dell'antigene:MAPT
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human,Rat,Mouse
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:4137
- Sinomini antigene:PPP1R103|FTDP-17|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Synthetic peptide
- Taglia:100 µl
- Conf:0,1 mL
Tau proteins are microtubule-associated proteins that are abundant in neurons in the central nervous system. In adult human brain, there are six tau isoforms that differ by the presence of either three or four C-terminal tandem repeated 31 or 33 amino acid sequences containing domains that are important for microtubule binding. Tau plays a crucial role in the neuron as it binds and stabilizes microtubules, and can regulate axonal transport; functions that are regulated by site-specific phosphorylation events. Abnormally hyperphosphorylated Tau protein, resulting in loss of function (the disruption of axonal transport) and gain of toxic function (formation of tau aggregates) at the same time, is deposited in cells as fibrillar lesions in numerous neurodegenerative diseases such as Alzheimer's disease, progressive supranuclear palsy, Pick's disease, corticobasal degeneration and post-encephalitic parkinsonism.
Western Blot: 1:2,000;
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat