Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:catenin (cadherin-associated protein), beta 1, 88kDa
- Simbolo dell'antigene:CTNNB1
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- ImmunoPrecipitazione:Yes
- Isotipo:IgG
- Reattività:Human,Rat,Mouse
- Western blot:Yes
- Epitopo:C-terminal
- Assorbimento incrociato:none
- ID gene:1499
- Sinomini antigene:armadillo|MRD19|CTNNB
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:His-tagged recombinant Human -Catenin (C-terminal fragment) protein purified from E. coli
- Purificazione:purified from E.coli
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
β-Catenin was a cytosolic protein originally identified through its association with the cadherin class of cell-adhesion proteins. β-Catenin has two key cellular functions; one plays direct role in cell adhesion/migration, bridging between cadherins and tactin cytoskeleton. The other plays as a transcription cofactor with T cell factor/lymphoid enhancer factor (TCF/LEF) in the Wnt pathway. Wnt are powerful regulators of cell proliferation and differentiation, and their signaling pathway involves proteins that directly participate in both gene transcription and cell adhesion. Activation of Wnt signaling leads to inhibition of Glycogen synthase-3β (GSK-3β) activity, resulting in accumulation of cytoplasmic (signaling) β-Catenin, which becomes available to bind the TCF/LEF family of transcription factors and to induce target gene expression. In the absence of Wnt signaling, A complex of axin and casein kinase-I(CK-I) induces β-Catenin phospho- rylation at a single site: serine 45(Ser45). This likely serves as a priming site for subsequent phosphorylation by GSK3b. And then β-Catenin is phosphorylated by GSK-3β at Ser33/37 and Thr41. These serine/threonine phosphylation is required for the phosph rylation-dependant degradation of β catenin via ubiquitin-proteosome pathway. Mutation of these phosphorylation sites in β-Catenin has been found in many tumor cell lines.
Western Blot: 1:2000; Immunoprecipitation: 1-2 ?l;
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat