Anti-LTF Rabbit Polyclonal Antibody
Lactoferrin(Lf), one of transferrin family, is found in external fluids, including milk and mucosal secretions, and prominent components of the secondary granules of neutrophils. Lf consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). Lf plays a central role in iron metabolism and host defense system against microbial infection.
Western Blot: 1:2000;
Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Taglia:100 µl
- Western blot:Yes
- Reattività:Human
- Assorbimento incrociato:none
- Immunogeno:Recombinant human protein purified from E.coli
- Purificazione:purified from E.coli
- Coniugazione:Non coniugato
- Isotipo:IgG
- Cat. No.:ABFRLF-PA0109
- Sinomini antigene:LF|HEL110|GIG12|HLF2
- Simbolo dell'antigene:LTF
- Clonalità:Policlonale
- Ospite:Rabbit
- ID gene:4057
- Temperatura di spedizione:Ice
- Tipo anticorpo:Primary
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Tipo di anticorpi:lactotransferrin
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



