Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Platelet-derived growth factor receptor beta
- Simbolo dell'antigene:PDGFRB
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human,Mouse
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:5159
- Sinomini antigene:JTK12|PDGFR|CD140B|IBGC4|PDGFR1|PDGFR-1|IMF1
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Synthetic peptide
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
Platelet-derived growth factors (PDGFs) have been implicated in the control of cell proliferation, survival and migration. The PDGF family of growth factors consists of five different disulphide-linked dimers built up of four different polypeptide chains encoded by four different genes. Theses isoforms, PDGFAA, PDGF-AB, PDGF-BB, PDGF-CC and PDGF-DD, act via two receptor tyrosine kinases, PDGF receptors α and β. Thus far, gene-targeting experiments have been attempted to create knockout mice deficient for PDGFR-α or PDGFR-β. Those mice, however, died either at the embryonic stage or several days after birth. Platelet-derived growth factor receptors, PDGFR-α and PDGFR-β, have five extracellular immunoglobulin-like domains and an intracellular tyrosine kinase domain. Upon binding a PDGF, the receptors form homo- and heterodimers. Dimerization of the receptors juxtaposes the intracellular part of the receptors, which allow phosphorylation in trans between the two receptors in the complex. These autophosphorylation provide docking sites for downstream signal transduction molecules. More than 10 different SH2– domaincontaining molecules have been shown to bind to different autophosphorylation sites in the PDGF α- and β-receptors. There are signal transduction molecules with enzymatic activity, such as PI3-kinase, PLC-γ, Src, SHP-2, GAP, as well as adaptor molecules such as Grb2, Shc, Nck, Grb7 and Crk, and Stats. Each of the different partners recruited by the activated receptor initiates different signaling pathways, making possible a great variety of cellular response.
Western Blot: 1:5,000;
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse