Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:cyclin D3
- Simbolo dell'antigene:CCND3
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human,Mouse
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:896
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Synthetic peptide
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
Cyclin D3, ~34kDa, is a member of cyclin D family that promotes cell cycle progression to the DNA systhesis (S) phase. Cyclins complexed with cyclin-dependent kinases(CDKs) mediate phosphorylation of other proteins relevant to cell proliferation. D-type cyclins and their complexing CDKs phosphorylate the retinoblastoma gene product with influences transcription of growth-controlling genes. Cyclin D3 regulates cell proliferation during hematopoiesis, carcinogenesis, and may have function in the terminally differentiated cells. In recent studies, there is reports that cyclin D3 involves in multiple myeloma and malignant precursor T cells.
Western Blot: 1:2,000;
Type: Primary
Antigen: CCND3 (cyclin D3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse