Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:peroxiredoxin 6
- Simbolo dell'antigene:PRDX6
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:9588
- Sinomini antigene:AOP2|NSGPx|p29|1-Cys|PRX|HEL-S-128m|aiPLA2
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Sulfonylated peptide(KLH coupled) corresponding to the active site sequence common to mammalian Prx VI
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
Peroxiredoxin VI (Prx VI, 1-Cys Prx) is a member of Peroxiredoxin Family, an antioxidant enzyme that detoxifies reactive oxygen species and has a cysteine at their active site. Six isoforms (Prx I to VI) of Prx exist in all eukaryotic cells. These isoforms are classified into three subgroups (2-Cys, atypical 2-Cys, and 1-Cys). Prx VI modulates various receptor-signaling pathways and protects cells from cell death induced by oxidative stress. The active site cysteine(Cys47) is oxidized to cysteine sulfenic acid(Cys47-SOH) by H2O2. However, the resulting Cys47-SOH does not form a disulfide bond because of unavailability of another Cys-SH nearby. It can be reduced by nonphysiological thiols such as DDT but is not transformed by Thioredoxin/Thioredoxin Reductase or GSH. Occasionally, the sulfenic intermediate is hyperoxidized to sulfinic or sulfonic acid, resulting in inactivation of peroxidase activity.
Western Blot: 1:2000;
Type: Primary
Antigen: PRDX6-SO3 (peroxiredoxin 6 (sulfonylated))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human