Order Entry
Switzerland
ContactUsLinkComponent
Anti-IFNA2 Mouse Monoclonal Antibody [clone: 24B10]
Anti-IFNA2 Mouse Monoclonal Antibody [clone: 24B10]
  ABFRLF-MA0364
Anti-IFNA2 Mouse Monoclonal Antibody [clone: 24B10]
  ABFRLF-MA0364

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Interferon alpha 2
  • Simbolo dell'antigene:
    IFNA2
  • Clonalità:
    Monoclonale
  • Clone:
    24B10
  • Coniugazione:
    Non coniugato
  • Ospite:
    Mouse
  • ImmunoPrecipitazione:
    Yes
  • Isotipo:
    IgG2b
  • Reattività:
    Human
  • Western blot:
    Yes
  • Assorbimento incrociato:
    none
  • ID gene:
    3440
  • Sinomini antigene:
    IFN-alphaA|INFA2|IFNA2B|IFNA
  • Temperatura di conservazione:
    Store for 1 year at –20 °C from date of shipment
  • Concentrazione:
    2 mg/ml
  • Temperatura di spedizione:
    Ice
  • Immunogeno:
    Recombinant human protein purified from E.coli (His-IFNa2)
  • Purificazione:
    Protein G purified
  • Taglia:
    100 µl
  • Conf:
    0,1 mL

 

 

Interferon alpha-2(IFNa2) which have antiviral activities, produced by macrophages. Three forms exist; alpha-2a, alpha-2b and alpha-2c.

Western Blot: 2.0 ?g/ml; Immunoprecipitation: 2.0 ?g;

Type: Primary
Antigen: IFNA2 (interferon, alpha 2)
Clonality: Monoclonal
Clone: 24B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human