Anti-COIL Mouse Monoclonal Antibody [clone: 1H10]
Catalog # ABFRLF-MA0358
Supplier: AbFrontier
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:coilin
- Simbolo dell'antigene:COIL
- Clonalità:Monoclonale
- Clone:1H10
- Coniugazione:Non coniugato
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG2b
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:8161
- Sinomini antigene:CLN80|p80-coilin
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (His-Coilin)
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
Coilin is a component of the nuclear Cajar(coiled) bodies (CBs) which are involved in the function or assembly/disassembly of nucleoplasmic snRNPs.
Human coilin is a 576-amino acid protein found enriched in CBs, but is also found in large amounts in the nucleoplasm.
Coilin is a constitutive phosphoprotein that is hyperphosphorylated during mitosis and it is thought that hyperphosphorylation triggers CB disassembly during cell replication. Self-interaction and localization have been shown to depend on the phosphorylation state of coilin
Western Blot: 0.5 ?g/ml; Immunoprecipitation: 2.0 ?g;
Type: Primary
Antigen: COIL (coilin)
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human