Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Dual specificity phosphatase 9
- Simbolo dell'antigene:DUSP9
- Clonalità:Monoclonale
- Clone:AF11B5
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG1
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:1852
- Sinomini antigene:MKP4|MKP-4
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (His-MKP4)
- Purificazione:Ammonium sulfate precipitation
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
There are ten mitogen-activated protein kinase (MAPK) phosphatases
(MKPs) that act as negative regulators of MAPK activity in mammalian
cells.
MKP4(DUSP9) is a dual specificity phosphatase, which acts as a negative
regulator of insulin-stimulated pathways. MKP4 is expressed in kidney,
placenta and adipose tissue. MKP4, a cytosolic MKP with specificity to not
only Erk, but also, to a lesser extent, JNK and p38. MKP4 also reversed
the effect of TNF-a to inhibit insulin Signaling. MKP4 has a protective
effect against the development of insulin resistance through its ability to
dephosphorylate and inactivate crucial mediators of stress-induced insulin
resistance.
Western Blot: 1:2,000;
Type: Primary
Antigen: DUSP9 (dual specificity phosphatase 9)
Clonality: Monoclonal
Clone: AF11B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human