- Tipo anticorpo:Primary
- Tipo di anticorpi:Insulin Receptor Substrate 2
- Simbolo dell'antigene:IRS2
- Clonalità:Monoclonale
- Clone:AF2F2
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG1
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:8660
- Sinomini antigene:IRS-2
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (His-IRS2)
- Purificazione:Ammonium sulfate precipitation
- Taglia:100 µl
- Conf:0,1 mL
Insulin receptor substrate (IRS) proteins play a central role in maintaining basic cellular functions such as growth and metabolism through insulin/insulin like growth factor (IGF) signaling. Four members (IRS-1, IRS-2, IRS-3, IRS-4) of this family have been identified which differ in their subcellular distribution and interaction with SH2 domain proteins. After phosphorylation by activated receptors, these intracellular signaling molecules recruit various downstream effector pathways including phosphatidylinositol 3-kinase, tyrosine protein phosphatase SHPTP-2, and several smaller adapter molecules such as the growth factor receptor-binding protein Grb-2.
IRSs are also thought to be able to induce malignant transformation.
Western Blot: 1:1,000;
Type: Primary
Antigen: IRS2 (insulin receptor substrate 2)
Clonality: Monoclonal
Clone: AF2F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human