Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Cyclin-dependent kinase inhibitor 1A
- Simbolo dell'antigene:CDKN1A
- Clonalità:Monoclonale
- Clone:AF9D10
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG1
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:1026
- Sinomini antigene:MDA-6|WAF1|P21|CAP20|p21CIP1|SDI1|CIP1|CDKN1
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (His-ABD-p21)
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
p21, also known as cyclin-dependent kinase inhibitor 1A or CDKN1A, directly inhibits the activity of cyclin-cdk2 and cyclin-cdk4 complexes. It mainly inhibits the activity of cyclin/cdk2 complexes and negatively modulates cell cycle progression. p21 also plays other fundamental roles including transcriptional regulation and the modulation of apoptosis.
p21 forms part of a family of structurally related proteins comprising p21, p27 and p57 that all share the ability to inhibit a wide range of cyclin -dependent kinase holoenzymes.
Besides its cdk inhibitor function, p21 is recognized to play a wide variety of physiological roles, many of which rely on its nuclear localization. p21 can bind to proliferating cell nuclear antigen (PCNA) thereby blocking DNA synthesis. p21 functions as a regulator of cell cycle progression at S phase. The expression of p21 is controlled by the tumor suppressor protein p53. Each of these functions of p21 is achieved through direct p21/protein interactions and the subcellular localization of p21 plays an important part in dictating the binding partners to which p21 is exposed.
Western Blot: 0.5 ?g/ml;
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: Monoclonal
Clone: AF9D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human