Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Nuclear factor of kappa light polypeptide gene enhancer in B cells 2 (p49/p100)
- Simbolo dell'antigene:NFKB2
- Clonalità:Monoclonale
- Clone:9D2
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG2b
- Reattività:Human,Rat,Mouse
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:4791
- Sinomini antigene:H2TF1|p52|LYT-10|p100|NF-kB2|CVID10|LYT10
- Numero amminoacido:1 to 454
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (His-NFkB 1-454 a.a.)
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
NF-κB (Nuclear Factor kappa B) is a nuclear transcription factor found in all cell types and is involved in cellular responses to stimuli such as stress, cytokines, free radicals, ultraviolet irradiation, and bacterial or viral antigens. NF-κB plays a key role in regulating the immune response to infection. Consistent with this role, incorrect regulation of NF-κB has been linked to cancer, inflammatory and autoimmune diseases, septic shock, viral infection and improper immune development. There are five members in the NF-κB family: NF-κB1, NF-κB2, RelA (also named p65), RelB, and c-Rel.
The most common form of NF-κB is the p50/RelA heterodimer, although other forms of NF-κB dimers, such as p50/p50, p52/p52, p52/RelA, p50/c-Rel, c-Rel/c-Rel, p52/RelB, and p50/RelB, have also been identified in some types of cells.
The primary role of NF-κB is to maintain normal cellular functions that range from cell-to-cell communication to cell motility, cell cycle progression, and cell lineage development. The activity of NF-κB is tightly regulated by interaction with inhibitory IκB proteins.
Western Blot: 0.5 ?g/ml;
Type: Primary
Antigen: NFKB2 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100))
Clonality: Monoclonal
Clone: 9D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat