Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Cyclin-dependent kinase 1
- Simbolo dell'antigene:CDK1
- Clonalità:Monoclonale
- Clone:6C9
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG2a
- Reattività:Human,Rat,Mouse
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:983
- Sinomini antigene:P34CDC2|CDC2|CDC28A
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (His-cdc2)
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
Cyclin-dependent kinases (cdk) belong to a group of protein kinases originally discovered as being involved in the regulation of the cell cycle. Cdks are also involved in the regulation of transcription and mRNA processing. A Cdk is activated by association with a cyclin, forming a cyclin-dependent kinase complex.
Cdk1, also known as cell division control protein 2 (cdc2), is one of the components of the maturation promoting factor (MPF) which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Cdk1, when bound to cyclin B, allows a dividing cell to enter into mitosis from G2 and permits the transition from G1 through S in conjunction with cyclin A and cyclin E. The Cdk1 protein is constantly present throughout the cell division cycle, but its activity is finely tuned by means of protein-protein interactions and reversible phosphorylation.
Cdk1 can also enhance cell migration. Increased levels of Cdk1 promote cell migration together with cyclin B2 and the actin-stabilizing protein caldesmon. Phosphorylation of caldesmon bound to actin results in the displacement of caldesmon from actin followed by the altered interaction of actin with myosin. These events contribute to increased cell migration.
Western Blot: 0.2 ?g/ml; Immunoprecipitation: 2.0 ?g;
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Monoclonal
Clone: 6C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat