Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Cyclin-dependent kinase 7
- Simbolo dell'antigene:CDK7
- Clonalità:Monoclonale
- Clone:3E3
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG2a
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:1022
- Sinomini antigene:CAK1|MO15|HCAK|CDKN7|STK1|p39MO15
- Numero amminoacido:136 to 346
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli(His/ABD-CDK7 136-346 a.a.)
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
Cyclin-dependent kinase 7 (CDK7) has essential roles in the cell-division cycle as a CDK-activating kinase (CAK) and in the transcription as a component of the general transcription factor TFIIH.
This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK), phosphorylating cell-cycle CDKs. It is also an essential component of the transcription factor TFIIH, which phosphorylates the C-terminal domain (CTD) of the largest subunit of Pol II. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle.
Embryonic stem cells (ESCs) shows a very unusual cell cycle structure, characterized by a short G1 phase and a high proportion of cells in S-phase. This is associated with a unique mechanism of cell cycle regulation by the activity of cyclin dependent protein kinase (Cdk). The unique cell cycle structure and mechanism of cell cycle control indicates that the cell cycle machinery plays a role in establishment or maintenance of the stem cell state.
Western Blot: 0.2 ?g/ml; Immunoprecipitation: 2.0 ?g;
Type: Primary
Antigen: CDK7 (cyclin-dependent kinase 7)
Clonality: Monoclonal
Clone: 3000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human