Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:zeta-chain (TCR) associated protein kinase 70kDa
- Simbolo dell'antigene:ZAP70
- Clonalità:Monoclonale
- Clone:49B4
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG1
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:7535
- Sinomini antigene:ZAP-70|SRK|TZK|STD|STCD
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli (ABD-ZAP70)
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
ζ-chain associated protein kinase, ZAP70, is a 70 kDa member of the Syk family kinase predominantly involved in T cell receptor (TCR) signaling. It is structurally homologous to Syk, a PTK that is involved in proximal BCR signaling. ZAP-70 is a key signaling molecule in T cell activation and also plays a role in apoptosis and cell migration.
SYK family tyrosine kinases contain a C-terminal kinase domain and tandem N-terminal SH2 domains that bind phosphorylated ITAMs (immunoreceptor tyrosine-based activation motif). Linker region that contains multiple tyrosines separates the SH2 domains from the kinase domain. Phosphorylated tyrosines act as docking sites for phospholipase Cγ1 (PLCγ1).
ZAP-70 and Syk are functionally homologous in antigen receptor signaling. Expression of ZAP-70 in Syk− B cells reconstitutes SCR function. Reconstitution requires the presence of functional Src homology 2 (SH2) and catalytic domains of ZAP-70.
Expression of ZAP-70 is an important negative prognostic factor in chronic lymphocytic leukemia (CLL) with more rapid disease progression and shorter survival.
Western Blot: 0.1ug/ml; Immunoprecipitation: 2.0ug;
Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: Monoclonal
Clone: 49B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human