Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:coagulation factor II (thrombin)
- Simbolo dell'antigene:F2
- Clonalità:Monoclonale
- Clone:28H5
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG2b
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:2147
- Sinomini antigene:PT|RPRGL2|THPH1
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Protein purified from Human plasma
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
Prothrombin precursor is synthesized in liver as a precursor of prothrombin containing a signal sequence and a propeptide domain at the N-terminus. Before its secretion into plasma, this precursor undergoes several posttranslational modifications, including removal of signal sequence and propeptide domains, glycosylation, and γ-glutamyl-carboxylation reaction. Prothrombin, cleaved by the prothrombinase enzyme complex that consists of serine protease factor Xa, cofactor Va, phospholipids and calcium, is converted to thrombin which converts fibrinogen into fibrin which in turn strengthens a protective clot. Activation of prothrombin is crucial in physiological and pathological coagulation. Various rare diseases involving prothrombin(e.g. hypoprothrombinemia) have been described.
Western Blot: 0.5 mg/ml;
Type: Primary
Antigen: Prothrombin precursor (prothrombin precursor)
Clonality: Monoclonal
Clone: 28H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human