Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:conserved helix-loop-helix ubiquitous kinase
- Simbolo dell'antigene:CHUK
- Clonalità:Monoclonale
- Clone:3A11
- Coniugazione:Non coniugato
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG1
- Reattività:Human,Mouse
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:1147
- Sinomini antigene:NFKBIKA|IKBKA|IKK-alpha|IKK1|IKKA|TCF16
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:2 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Synthetic peptide
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
IκB kinase α (IKKα) is a component of a multiprotein kinase complex that regulates the activity of the transcription factor NF-κB. Activation of the IKK complex via upstream stimuli leads to phosphorylation and degradation of NF-κB bound IκB(Inhibitor of κB). Subsequently, free NF-κB dimers enter the nucleus and regulate the transcription of a variety of target genes involved in cell proliferation and differentiation, apoptosis, and inflammation, etc. The IKK complex consists of 2 highly homologous kinase subunits, IKKα and IKKβ, and a nonenzymatic regulatory component, IKKγ/NEMO. The relative contributions of IKKα and IKKβ to the signalling complex vary according to the requirements of the cell. IKKβ plays a predominant role in immune responses, while IKKα alone appears to be sufficient for at least some developmental systems. Recently, IKK/NF-κB signaling pathway was studied as therapeutic targets in cancer.
Western Blot: 2.0 ?g/ml; Immunoprecipitation: 2 ?g;
Type: Primary
Antigen: CHUK (conserved helix-loop-helix ubiquitous kinase)
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat