Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Haptoglobin
- Simbolo dell'antigene:HP
- Clonalità:Monoclonale
- Clone:26E11
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG1
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:3240
- Sinomini antigene:HPA1S|BP|HP2ALPHA2
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Protein purified from Human plasma
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
Haptoglobin (abbreviated as Hp) is a protein in the blood plasma that binds free hemoglobin released from erythrocytes with high affinity and thereby inhibits its oxidative activity. Hp in its simplest form consists of two α- and two β-chains, connected by disulfide bridges. The chains originate from a common precursor protein which is proteolytically cleaved during protein synthesis. Hp exists in two allelic forms in the human population, so called Hp1 and Hp2; the latter one having arisen due to the partial duplication of Hp1 gene. Three phenotypes of Hp are found in humans: Hp1-1, Hp2-1, and Hp2-2. Hp phenotypes are associated with pathogenesis of a number of human disorders, such as diabetes, cardiovascular disease, etc. Hp plays a role in the host defence responses to infection and inflammation, acting as a natural antagonist for receptor-ligand activation of the immune system, also.
Western Blot: 1.0 ?g/ml;
Type: Primary
Antigen: HP (haptoglobin)
Clonality: Monoclonal
Clone: 2600000000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human