Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Mitogen-activated protein kinase 3
- Simbolo dell'antigene:MAPK3
- Clonalità:Monoclonale
- Clone:9B3
- Coniugazione:Non coniugato
- Ospite:Mouse
- Isotipo:IgG2b
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:5595
- Sinomini antigene:HS44KDAP|ERK1|HUMKER1A|p44-MAPK|ERT2|PRKM3|P44ERK1|P44MAPK|ERK-1|p44-ERK1
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways.
When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase(ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.
Western Blot: 0.2 ?g/ml;
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: Monoclonal
Clone: 9B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat