Anti-LTF Mouse Monoclonal Antibody [clone: 1B8]
Codice catalogo ABFRLF-MA0109
Nome del fornitore: AbFrontier
Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:lactotransferrin
- Simbolo dell'antigene:LTF
- Clonalità:Monoclonale
- Clone:1B8
- Coniugazione:Non coniugato
- Ospite:Mouse
- ImmunoChimica:Yes
- ImmunoPrecipitazione:Yes
- Isotipo:IgG1
- Reattività:Human
- Assorbimento incrociato:none
- ID gene:4057
- Sinomini antigene:LF|HEL110|GIG12|HLF2
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from yeast pichia pastoris
- Purificazione:Ammonium sulfate precipitation
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
Lactoferrin(Lf), one of transferrin family, is found in external fluids, including milk and mucosal secretions, and prominent components of the secondary granules of neutrophils. Lf consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). Lf plays a central role in iron metabolism and host defense system against microbial infection.
Immunoprecipitation: 1~2 ?l; Immunohistochemistry: Paraffin-embedded sections;
Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human