Anti-Ubiquitin +1 Mouse Monoclonal Antibody [clone: 40B3]
ABFRLF-MA0087
- Tipo anticorpo:Primary
- Tipo di anticorpi:Ubiquitin A-52 Residue Ribosomal Protein Fusion product 1
- Simbolo dell'antigene:UBA52
- Clonalità:Monoclonale
- Clone:40B3
- Coniugazione:Non coniugato
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG1
- Reattività:Human
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:7311
- Sinomini antigene:RPL40|CEP52|L40|HUBCEP52
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Synthetic peptide (KLH coupled) corresponding to C-terminal residues of human Ubiquitin +1.
- Purificazione:Ammonium sulfate precipitation
- Taglia:100 µl
- Conf:0,1 mL
Ubiquitin+1 (Ub+1) is a novel mutant form of ubiquitin that can be produced through a process known as molecular frameshift. Ub+1 can be polyubiquitinated to produce aberrant polyubiquitin chains that inhibit the 26S proteasome. Especially ub+1 is accumulated in aggregates containing amyloid-β and phosphorylated-tau. Elevated expression of Ub+1 mRNA and protein has been observed in the brains of patients with Alzheimer's disease. Also ub +1 acts as an aggravating factor in polyglutamine-induced neuro-degeneration.
Western Blot: 1:500-1000; Immunoprecipitation: 1-2 ?l;
Type: Primary
Antigen: Ubiquitin +1 (ubiquitin +1)
Clonality: Monoclonal
Clone: 40B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human