Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Thioredoxin 2
- Simbolo dell'antigene:TXN2
- Clonalità:Monoclonale
- Clone:4C5
- Coniugazione:Non coniugato
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG2b
- Reattività:Human,Rat,Mouse
- Assorbimento incrociato:none
- ID gene:25828
- Sinomini antigene:TRX2|MTRX|MT-TRX
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli
- Purificazione:Ammonium sulfate precipitation
- Taglia:100 µl
- Conf:0,1 mL
Specifications
About this item
Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/ dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cysteine residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds (1). Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations (2). Specific protein disulfide targets for reduction by Trx include protein disulfide–isomerase (PDI) (3) and a number of transcription factors such as p53 (4), NF-kB (5) and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins (6-7).
Immunoprecipitation: 2 ?l;
Type: Primary
Antigen: TXN2 (thioredoxin 2)
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat