- Tipo anticorpo:Primary
- Tipo di anticorpi:v-myc avian myelocytomatosis viral oncogene homolog
- Simbolo dell'antigene:MYC
- Clonalità:Monoclonale
- Clone:LF10E3
- Coniugazione:Non coniugato
- Ospite:Mouse
- ImmunoPrecipitazione:Yes
- Isotipo:IgG1
- Western blot:Yes
- Assorbimento incrociato:none
- ID gene:4609
- Sinomini antigene:MRTL|bHLHe39|c-Myc|MYCC
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Concentrazione:1 mg/ml
- Temperatura di spedizione:Ice
- Immunogeno:synthetic peptide derived
- Purificazione:Protein G purified
- Taglia:100 µl
- Conf:0,1 mL
Epitope tags can be used to label and detect proteins with immunoblotting, immunoprecipitation and immunostaining techniques.
Due to their small size, they are unlikely to affect the tagged proteina's biochemical properties. The Myc epitope tag is widely used to detect
expression of recombinant proteins in bacteria, yeast, insect and mammalian cell systems. Myc proteins are nuclear proteins with relatively
short half-lives. Amplification of the c-Myc gene has been found in several types of human tumors including lung, breast and colon carcinomas.
Western Blot: 0.1 ?g/ml; Immunoprecipitation: 2.0 ?g;
Type: Tag
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: Monoclonal
Clone: LF10E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: