Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Glutathione reductase
- Simbolo dell'antigene:GSR
- Clonalità:Monoclonale
- Clone:1A7
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- ImmunoChimica:Yes
- ImmunoPrecipitazione:Yes
- Isotipo:IgG2a
- Reattività:Human
- Assorbimento incrociato:none
- ID gene:2936
- Sinomini antigene:HEL-75
- Temperatura di conservazione:Store for 1 year at –20 °C from date of shipment
- Temperatura di spedizione:Ice
- Immunogeno:Recombinant human protein purified from E.coli
- Purificazione:Ammonium sulfate precipitation
- Taglia:100 µl
- Conf:0,1 mL
Specifiche
Informazioni su questo articolo
Glutathione reductase (GR) is a member of pyridine nucleotide-disulfide oxidoreductases, which includes the closely related enzymes thioredoxin reductase, lipoamide dehydrogenase, trypanothione reductase and mercuric ion reductase. GR is a cytoplasmic flavoenzyme widely distributed in aerobic organisms. The dimeric protein is composed of two identical subunits, each containing 1 FAD and 1 redox-active disulfide/dithiol as components of the catalytic apparatus. It plays a role in maintaining glutathione (GSH) in its reduced form by catalyzing the reduction of glutathione disulfide (GSSG) (1):
GSSG + NADPH + H+ 2GSH + NADP+
In most eukaryotic cells, GR maintains the ratio of [GSH]/[GSSG] elevated, and participates in several vital functions such as the detoxification of reactive oxygen species as well as protein and DNA biosynthesis (2).
Immunoprecipitation: 1-2 ?l; Immunocytochemistry: 1:100;
Type: Primary
Antigen: GSR (glutathione reductase)
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human