Scraper, SP Bel-Art
286 mm, 140×63 mm
Catalog # BELAH368200000
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
Polyamide, glass-reinforced. Rigid and strong one piece scraper for heavy use.
- Flat blade with chisel edge
- Blade may be resharpened to restore chisel edge
Specifications
- Appl. area di pulizia:140×63 mm
- Lunghezza:286 mm
- Seq:02