Peqlab, PCR Plates, 96-Well, Non-Skirted
Nome del fornitore: VWR Collection
Certificati
Informazioni su questo articolo
PP, non-skirted. Standard 96-well PCR plates, suitable for a wide range of 0,2 ml thermal cycler blocks.
- Effective design with overhanging edges for easy handling
- Cut corner (H1)
- Can be sealed using Thermo-Mats™, adhesive films and foils, heat seals and flat or domed cap strips
- White plates for Real-Time PCR
- Certified free from DNase, RNase and human DNA
Specifiche
- Catalog No:
- 732-2879
- 732-3214
- Environmentally Preferable:
- Produzione a basso impatto
- Produzione a basso impatto
- Certificazione ambientale:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)