Compression dressing, Dermotekt®
Catalog # SOEH1003401
Supplier: SOEHNGEN
Specifications
- Descrizione:DERMOTEKT® compression dressing made from non-woven polyester, sterile, in peel-off packaging
- L×L:100×100 mm
- Conf:2
Specifications
About this item
Dressing made from high-volume, non-woven polyester with a high absorbency of 1000 g/m².
- Smooth surfaces, no adherence to wounds, no wicking action
- Physiologically harmless, breathable, supple
- No chemical binding agents or optical brighteners
- Grey/red colour code
- EO-sterilised
Certifications: Exceeds DIN 13152 requirements.