SP Bel-Art® liquid sampler
Catalog # BELAH378270000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacità:50 ml
- Descrizione:Liquid sampler
- Lunghezza:500 mm
- Materiale:Ottone nichelato
- Conf:1
Specifications
About this item
Liquid sampler made of nickel-plated brass cup and tubular handle with finger grip. Cup is not replaceable and can not detach.
Suitable for immersion into a can, bottle, tank, vat ...