Deionisers, labwater
Deioniser, Labwater 2
Codice catalogo PURIL991020
Nome del fornitore: Purite
Vai a
Product Details & Documents
These deionisers are offer a simple, cost effective method of producing 1 to 10 μS/cm purified deionised water at low volumes. Typically these units are ideal where water usage is up to 10 litres per day.
- Incorporate easily replaceable cartridge, containing specifically designed resin, which changes colour through absorption of ionic contaminants, facilitating a low level of maintenance
Available in two versions offering maximum flow rates of 30 and 60 litres per hour and complete with: wall brackets and connections, deionisation cartridge plus one spare, colour change resin, inlet on/off valve with 12 mm hose connector and outlet spout with flexible pipe
: The units are typically wall mounted and are simply connected via a dedicated hose to a tap or stopcock and operate direct from water pressure thus requiring no external electrical supply.
Specifiche
- Descrizione:Deioniser, Labwater 2
- Seq:02
- Cat. No.:PURIL991020
- Peso:4,4 kg
- Modello:Labwater 2
- Pressione di esercizio max.:5 bar
- Portata di flusso max.:60 l/h
- L×P×A:80×100×760 mm
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement cartridge for Labwater 1
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement cartridge for Labwater 2