Anion exchange columns, Dionex™, IonPac™ AS9-HC
Supplier: DIONEX
Certificates
About this item
The IonPac™ AS9-HC provides baseline separation of chlorite, chlorate, bromate, and bromide from common anions and provides good resolution between bromate and chloride to allow a low detection level of 5 µg/L (5 ppb) for bromate in the presence of a high chloride (50 mg/L) concentration. Specifically designed for trace bromate in drinking water.
Frequently Bought Together