Transfer pipettes, Transpette™, SP Bel-Art
Codice catalogo 710-0083
Nome del fornitore: Bel-Art Products, a Part of SP
Specifiche
- Confezione:Individually wrapped, 100/box
- Sterili:+
- Capacità:8×600 µl
- Seq:02
- Conf:100
Specifiche
Informazioni su questo articolo
Economical, single-use, 8-channel transfer pipette made of translucent plastic. Ideal for washing plates, feeding and supernatant aspiration from microwell plates.
- No expensive pipettes and pipette tips needed
- Should be disposed directly after 'hot' uses in order to avoid radiation contamination of costly pipettes
- Available sterile or non sterile; can't be steam sterilised
- Capacity: 600 µl per channel