Multipurpose trays, with lid, SP Bel-Art
Codice catalogo BELAF162300000
Nome del fornitore: Bel-Art Products, a Part of SP
Specifiche
- L×P×A:203×273×76 mm
- Conf:1
Specifiche
Informazioni su questo articolo
PP.
- Chemical and corrosion resistant
- Flexible and strong
- Stackable to conserve space
Multipurpose trays with snap on lid is ideal for safely storing small items and holding instruments while autoclaving. Rounded and smooth inside corners allow easy cleaning.