Orders LinkContactUsLinkComponent
ICP multi-element standards
ICP multi-element standards
Nome del fornitore:  AGILENT HEWLETT PACKARD
Certificates
ICP multi-element standards
ICP multi-element standards
4 varianti
Product Details & Documents
Product Family Options
Informazioni sul prodotto
  • Concentrazione
    Matrice
    Conf
    Disponibilità
    Price
  • HEWL8500-6940
    Multi-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn
    Multi-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn
    10 mg/L
    5% HNO₃
    1
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HEWL8500-6942
    Multi-element calibration standard-4, 100 ml, (12 elements in H20/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr
    Multi-element calibration standard-4, 100 ml, (12 elements in H20/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr
    10 mg/L
    1% HNO₃ + (CHOH)₂(COOH)₂
    1
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HEWL8500-6944
    Multi-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb
    Multi-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb
    10 mg/L
    5% HNO₃
    1
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HEWL8500-6948
    Multi-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn
    Multi-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn
    10 mg/L
    10% HCl
    1
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..