Pens
Spare recording pen, viola
Codice catalogo THIE500847
Nome del fornitore: Thies Clima
Vai a
Product Details & Documents
Felt pen for recording instruments, like Hygrograph and Hygrothermograph.
Specifiche
- Colore:viola
- Descrizione:Spare recording pen