Shell vial convenience kits with SepCap™
Supplier: THERMO NATIONAL SCIENTIFIC
Certificates
About this item
Frequently Bought Together
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
FILTER VIAL 0.45uM PVDF, PRE-SLIT CAP 1 * 500 ST
Supplier: THOMSON INSTRUMENT
Avantor® ACE® C18-AR, colonne HPLC, 3 µm
Supplier: Avantor
L-MARK® Replacement Plunger for CTC Analytics PAL DLW (Dynamic Load and Wash)
Supplier: Trajan Scientific and Medical
[EN]COLUMN CHIRALPAK IM 250X4.6MM 5M 1 * 1 pezzi
Supplier: CHIRAL TECHNOLOGIES
[EN]NARROW NECK BOTTLE 2000ML WITHOUT CO 1 * 1 pezzi
Supplier: PerkinElmer
Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
HPLC columns, Hypersil™ BDS C8
Supplier: Thermo Fisher Scientific
Vydac 214TP C4, HPLC columns
Supplier: Avantor
Waste tubes, b.safe
Supplier: Bohlender
Chromatography Tubing, Stainless Steel
Supplier: Avantor
Electronic crimping tools
Supplier: MACHEREY-NAGEL