Cuvette washer, Vakuwash®, SP Bel-Art
Catalog # BELAF389600000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Descrizione:Cuvette washer with PP vacuum flask, vinyl stopper and PVC tubing for cuvettes of I.Ø ≥3 mm (funnel Ø = 89 mm)
- Conf:1
Specifications
About this item
Economical, durable and gentle all-plastic washer for delicate glass or quartz cuvettes.
- Mouth of an inverted cuvette is pressed against the rubber cushion to induce suction
- Water or other cleaning fluid poured into the 89 mm (3½") Ø funnel is forcibly sprayed into the cuvette and then drains into the flask
- Continued pressure on the cuvette creates an air flow which dries it
- Final wash in acetone aids in removing water droplets
- Minimum cuvette Int.Ø must be 9 mm or larger to form a seal over the spray nozzle
As long as the cuvette maintains pressure against the cushion, the continued force sets up a stream of air flow into the cuvette and drys it thoroughly. Any residual water droplets may be removed with a final rinse with acetone.
Delivery: Supplied with 500 ml PP vacuum flask, Vikem® vinyl stopper and 3' PVC tubing ready for connection to a vacuum line.