Washed pooled cells, human blood, non sterile
Anbieter: Rockland Immunochemicals
Zertifikate
Über diesen Artikel
Human whole blood is washed to remove the platelet rich plasma, buffy coat layer and leukocytes (WBC). It is used for complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents. Store cells at 4 °C prior to opening.
Frequently Bought Together