Manganese reagent set, LR
Catalog # HACH2243300
Supplier: Hach
CAS Number:
Specifications
- Metodo chimico:PAN
- Conf:100 Tests
- Descrizione:Manganese reagent set
- Range di misura:0,006 - 0,700 mg/l Mn
Specifications
About this item
Reagent set used for determination of manganese by the PAN method.
Delivery: Reagent set includes ascorbic acid powder pillows, alkaline cyanide reagent and PAN indicator solution (0,1% for 100 tests).