Nitrite test kit, colour disc, NI-6
Catalog # HACH224000
Supplier: Hach
CAS Number:
Specifications
- Conf:1 KIT
- Descrizione:Nitrite test kit, HR, NI-6
- Range di misura:0 - 80 mg/l NO₂
0 - 1600 mg/l NO₂
Specifications
About this item
The kit uses the colour disc or ferrous sulphate method to measure nitrogen as nitrite NO²⁻.
- Simple, cost effective
- Portable test kit for analysis in the field
Delivery: Kit includes colour disc, comparator box and tubes, reagents, deionised dilution water, instructions and carrying case.