Water quality test discs, Model HA-77
HACH202300
: Hach
:
- Conf:1 KIT
- Descrizione:Hardness and iron color disc test kit, Model HA-77
- Range di misura:0 - 5 mg/l
Hardness and iron determination in a water quality test kit which offers accuracy and portable utility in one package. Utilising drop count titration (hardness) and colour disc colorimetry.
- Easy to use
- Portable kit
Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
: Includes four reagents, bottles, flask, color discs, comparator box, viewing tubes, instructions and carrying case sufficient for 100 tests.