Lauryl tryptose broth MPN tubes
Catalog # HACH2217515
Anbieter: Hach
CAS-Nummer:
Technische Daten
- Conf:15
- Standard:-
- Descrizione:Lauryl tryptose broth MPN tubes, single strength
- Volume:15 ml
Technische Daten
Über diesen Artikel
Lauryl tryptose broth tubes with MUG microbiological media, single strength.
- Save time and labour costs
- Ready to use MPN tubes
- With minimal training requirements