Blocking reagents, Blotto
Blocking reagent, Blotto, immunoanalytical grade, 500 g
Nome del fornitore: Rockland Immunochemicals
Blocking reagents, Blotto
Codice catalogo ROCKB501-0500
Nome del fornitore: Rockland Immunochemicals
Numero CAS:
Vai a
Product Details & Documents
Contains immunoanalytical grade non-fat dry milk. Blocks non specific binding of proteins in various immunological assays. Ideal for use in western blotting and ELISA methods.
Specifiche
Specifiche
- Conf:500 g
- Descrizione:Blocking reagent, Blotto, immunoanalytical grade