Radtape™ labels
Radtape labels, nero, 102×152,4 mm
Codice catalogo DIVERAD-10
Nome del fornitore: DIVERSIFIED BIOTECH
Vai a
Product Details & Documents
These RadTape™ labels are used to identify autograms. They are phosphorescent, non-radioactive adhesive labels that may be cut to the specific size and shape needed for each individual membrane, dried gel or microarray.
- Ideal for chemilluminescence and radiological autography
- Great for documentation and archiving
- Use your own codes and symbols
- Fast and easy marking system
An ordinary pen may be used to mark the label using the investigator’s own codes and symbols. The custom label may then be peeled from its backing and affixed to the membrane, gel or microarray. Once placed against film, Radtape™ exposes a negative image of the markings directly onto the autorad, thus producing a permanent record of the experiment.
Specifiche
- Colore:nero
- Descrizione:Radtape labels
- Dimensioni:102×152,4 mm