PCR plates, Eppendorf twin.tec® LoBind
Catalog # EPPE0030129.571
Supplier: EPPENDORF
Specifications
- Colore del telaio:arancione
- N. di pozzetti:96
- Profilo:Standard
- Bordo:Fully skirted
- Colore del pozzetto:trasparente
- Seq:10
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:Renewable Energy Certificate, ISO 14001
- Conf:300
- Volume pozzetti:0,15 ml
Specifications
About this item
Eppendorf twin.tec® PCR plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and robust polycarbonate frames for ultimate rigidity and torque-resistance. Eppendorf twin.tec® plates reduce the risk of cross contamination with raised well rims for effective sealing.
- PCR clean
These Eppendorf twin.tec® 96 plates are ideal for quantitative Real-Time PCR, standard PCR, high-throughput PCR and small volume sample handling.
Ordering information: PCR plates are available with barcode upon request, please contact your local Avantor sales office for further details.