Clamps for standard taper joints, WHEATON®
Catalog # WHEA297755
Supplier: DWK Life Sciences
Specifications
- per cono:29
- Colore:rosso
- Seq:02
- Conf:12
Specifications
About this item
These colour-coded clamps moulded from polyoxymethylene (POM), an acetal resin, hold standard taper joints securely under moderate pressure.
- Will not scratch glass and can withstand temperatures up to 150 °C
- Resistant to bases and weak acids
- Not resistant to halogen gases and concentrated acids